missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Fetal Bovine Serum, Premium, New Zealand Origin
Collected at government registered facilities in New Zealand
1365.00 €
Specifications
Content And Storage | -40°C to -10°C |
---|---|
Species | Bovine |
Country of Origin | New Zealand |
Form | Liquid |
Sterility | Sterile |
Description
- Exported from the country of origin accompanied with a Veterinarian Health Certificate and imported under a USDA-APHIS issued import permit.
- True pooled and is triple 0.1 micron sterile filtered that passes sterility, mycoplasma, and 9CFR viral testing requirements.
- Validated process used to heat inactivate serum at 56°C ± 2°C for 30 minutes via bulk inactivation of entire lot in the pooling tank or individual bottles in a water bath.
Specifications
-40°C to -10°C | |
New Zealand | |
Sterile |
Bovine | |
Liquid | |
New Zealand Origin |
Safety and Handling
missing translation for 'shelfLife' : 60 months
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title