Learn More
Gibco™ Fetal Bovine Serum, mesenchymal stem cell-qualified, USDA-approved regions
Gibco™ fetal bovine sera is specially tested for the ability to support the expansion and clonal enumeration (colony forming unit fibroblast assay) of mesenchymal stem cells.
Brand: Gibco™ 12662011
Description
Gibco™ fetal bovine sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements
- Sera Category: Specialty
- Origin: Countries meeting USDA importation requirements
- Endotoxin level: ≤50 EU/mL (levels routinely ≤10 EU/mL)
- Hemoglobin level: ≤25 mg/dL
- Storage: Store at -5°C to -20°C.
Specially tested for the ability to support the expansion and clonal enumeration (colony forming unit fibroblast assay) of mesenchymal stem cells.
Order Info
Shipping Condition: Dry Ice
Specifications
Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
Cattle/Bovine | |
Liquid | |
Fetal Bovine Serum | |
Standard (Sterile-filtered) | |
<50 EU/mL | |
MS Cell-Qualified |
Fetal | |
Countries meeting USDA importation requirements | |
Bottle | |
100 mL | |
Sterile | |
≤25 mg/dL | |
Frozen |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.