Learn More
Thermo Scientific™ Hin1II (NlaIII) (5 U/μL)
Cut at CATG^ sites with Hin1II (NlaIII) restriction enzyme, which performs best at 37°C in G buffer (Isoschizomers: CviAII, FaeI, FatI, Hsp92II, NlaIII).
Brand: Thermo Scientific™ ER1831
43.20 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
Lambda DNA digested with Hin1II (NlaIII), 1.4% agarose, 181 cleavage sites
conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
Highlights
- Superior quality -stringent quality control and industry leading manufacturing process
- Convenient color-coded Five Buffer System
- Includes universal Tango buffer for double-digestions
- BSA premixed in reaction buffers
- Wide selection of restriction endonuclease specificities

Specifications
5 U/μL | |
Hin1II (NlaIII) | |
Yes | |
No | |
Restriction Enzyme |
Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive | |
10X Buffer G | |
37°C | |
300 units | |
Traditional Cloning |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.