Learn More
Thermo Scientific™ Acc65I (Asp718I) (10 U/μL)
Cut at G^GTACC sites with Acc65I (Asp718I) restriction enzyme, which performs best at 37°C in O buffer (Isoschizomers: Asp718I).
59.50 € - 238.00 €
Specifications
Concentration 10 U/μL
Methylation Sensitivity CpG Methylation-Sensitive, Not CpG Methylation-Sensitive, Not Dam Methylation-Sensitive
Enzyme Acc65I (Asp718I)
Compatible Buffer 10X Buffer O
Sensitive to Heat Inactivation Yes
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10232680
View Documents
Thermo Scientific™
ER0901
1,000 units
59.50 €
1000 units
10713911
View Documents
Thermo Scientific™
ER0902
5,000 units
238.00 €
5000 units
Description
Description
Conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
Highlights
  • Superior quality -stringent quality control and industry leading manufacturing process
  • Convenient color-coded Five Buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities
Specifications
Specifications
10 U/μL
Acc65I (Asp718I)
Yes
No
Traditional Cloning
CpG Methylation-Sensitive, Not CpG Methylation-Sensitive, Not Dam Methylation-Sensitive
10X Buffer O
37°C
Restriction Enzyme