Learn More
Hellma™ Far UV Quartz Matched Pairs Macro Cells
Brand:  Hellma™ 100-5-K-44E
Product Code. 12741715
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Specifications
Matched Pairs Macro Cell
Far UV Quartz
5 mm
1.75 mL
2Pack
200 to 2500 nm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hellma™ Far UV Quartz Matched Pairs Macro Cells > Capacity: 1.75mL