Learn More
FANCL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14454-1-AP-20UL
 View more versions of this product
Product Code. 16853104
  • 145.00 € / 20µL
Explore more special offers
Description
Description
The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ (also called BRIP1), FANCL, FANCM and FANCN (also called PALB2). The previously defined group FANCH is the same as FANCA. Fanconi anemia is a genetically heterogeneous recessive disorder characterized by cytogenetic instability, hypersensitivity to DNA crosslinking agents, increased chromosomal breakage, and defective DNA repair. The members of the Fanconi anemia complementation group do not share sequence similarity; they are related by their assembly into a common nuclear protein complex. This gene encodes the protein for complementation group L. Alternative splicing results in two transcript variants encoding different isoforms.
Specifications
Specifications
FANCL
Polyclonal
Unconjugated
FANCL
FAAP43, FANCL, Fanconi anemia group L protein, PHF9, POG
Rabbit
Antigen Affinity Chromatography
RUO
305600, 55120, 67030
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9CR14, Q9NW38
FANCL
FANCL Fusion Protein Ag6031
20 μL
Primary
Mouse, Human, Rat