missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FANCL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14454-1-AP-150UL
This item is not returnable.
View return policy
Description
The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ (also called BRIP1), FANCL, FANCM and FANCN (also called PALB2). The previously defined group FANCH is the same as FANCA. Fanconi anemia is a genetically heterogeneous recessive disorder characterized by cytogenetic instability, hypersensitivity to DNA crosslinking agents, increased chromosomal breakage, and defective DNA repair. The members of the Fanconi anemia complementation group do not share sequence similarity; they are related by their assembly into a common nuclear protein complex. This gene encodes the protein for complementation group L. Alternative splicing results in two transcript variants encoding different isoforms.Specifications
FANCL | |
Polyclonal | |
Unconjugated | |
FANCL | |
FAAP43, FANCL, Fanconi anemia group L protein, PHF9, POG | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
305600, 55120, 67030 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9CR14, Q9NW38 | |
FANCL | |
FANCL Fusion Protein Ag6031 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |