missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM65B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17015-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody is specific to the isoform of 119 kDa of FAM65B.
Required for hearing. Involved in skeletal muscle development.Specifications
FAM65B | |
Polyclonal | |
Unconjugated | |
RIPOR2 | |
C6orf32, DIFF40, DIFF48, FAM65B, KIAA0386, PL48, Protein FAM65B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
193385, 306934, 9750 | |
-20°C | |
Liquid |
Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q7TP54, Q80U16, Q9Y4F9 | |
RIPOR2 | |
FAM65B Fusion Protein Ag9991 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction