missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM127B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20282-1-AP-20UL
This item is not returnable.
View return policy
Description
The function of this protein remains unknown.Specifications
FAM127B | |
Polyclonal | |
Unconjugated | |
RTL8A | |
CXX1b, FAM127B, MAR8A, Protein FAM127B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26071, 66158, 679038 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.36 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BWD3 | |
Rtl8a | |
FAM127B Fusion Protein Ag13930 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction