Learn More
FAM127B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20282-1-AP-150UL
 View more versions of this product
Product Code. 16869804
  • 410.00 € / 150µL
Explore more special offers
Description
Description
The function of this protein remains unknown.
Specifications
Specifications
FAM127B
Polyclonal
Unconjugated
RTL8A
CXX1b, FAM127B, MAR8A, Protein FAM127B
Rabbit
Antigen Affinity Chromatography
RUO
26071, 66158, 679038
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.36 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BWD3
Rtl8a
FAM127B Fusion Protein Ag13930
150 μL
Primary
Human, Mouse, Rat