missing translation for 'onlineSavingsMsg'
Learn More
Learn More
F2RL3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25306-1-AP-150UL
This item is not returnable.
View return policy
Description
Coagulation factor II (thrombin) receptor-like 3 (F2RL3) is a member of the large family of 7-transmembrane-region receptors that couple to guanosine-nucleotide-binding proteins. F2RL3 is also a member of the protease-activated receptor family. F2RL3 is activated by proteolytic cleavage of its extracellular amino terminus. The new amino terminus functions as a tethered ligand and activates the receptor. F2RL3 is activated by thrombin and trypsin.Specifications
F2RL3 | |
Polyclonal | |
Unconjugated | |
F2RL3 | |
F2RL3, PAR 4, PAR4, Proteinase activated receptor 4, Thrombin receptor like 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14065, 9002 | |
-20°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O88634, Q96RI0 | |
F2RL3 | |
F2RL3 Fusion Protein Ag20801 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |