Learn More
Julabo™ TopTech Series Refrigerated/Heating Circulating Baths with ME Circulators
For working temperatures from -50deg.C to +200°C. Julabo™ TopTech Series Refrigerated/Heating Circulating Baths with ME Circulators has connection for external Pt100 sensor for direct measuring and control in external application.
Brand:  Julabo™ 9162634
 View more versions of this product
Product Code. 11934818
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • VFD Comfort Display
  • PID3 temperature control, stability ±0.01°C
  • Keypad for setpoints, warning/safety values and menu functions
  • Integrated programmer (1 x 10 steps) with real-time clock
  • ATC3 3-Point-Calibration
  • Pt100 External sensor connection for measurement and control
  • SMART PUMP, electronically adjustable pump stages
  • Adjustable high temperature cut-out, visible via display
  • Early warning system for low liquid level (DBGM 203 06 059.8)
  • RS232 interface for online communication
  • Integrated programmer for 10 program steps
  • Active Cooling Control
Specifications
Specifications
PID, Cascade
Classification III (FL) according to DIN 12876-1
11 to 16 L/min.
VFD
2 barbed fittings suitable for tubing 8 and 10mm inner dia. (pump connections M10x1 female)
F34-ME
58 x 38 x 62 cm
23.8 cm
14.9 cm
42 kg
115 V
60 Hz
Interior (L x W x D): 30 x 24 x 15cm
0.45kW at 20°C, 0.32kW at 0°C, 0.14kW at -20°C
5°C to 40°C
1 kW
0.23 to 0.45 bar
±0.01°C
38.1 cm
61.9 cm
41.7 kg
115 V 60 Hz 14 A
14 A