missing translation for 'onlineSavingsMsg'
Learn More

Manutan Extra shelf for Rapid 2 medium duty shelving, grey, 915mm x 455mm

Brand:  Manutan A364257

Encompass_Preferred

Product Code. 12888148

  • 53.72 € / Each
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Economical chipboard suited for everyday applications. Select the same dimensions (W x D, mm) as the Shelving Bays for the correct size required. Grey beams and chipboard shelf cover, 16mm high density chipboard.
  • Grey
  • Shelf capacity UDL: 340kg
  • Dimensions (W x D): 915mm x 455mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers