missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius™ External Adjusting Weight With DKD Calibration Certificate
Test weight weight set for special weighing instruments
Brand: Sartorius™ YSS43-02
269.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Calibration weight 30g ± 0.3mg
- DAkkS Certificate
- Special, non-magnetizable steel

Specifications
DAkkS Certificate | |
Calibration weight | |
Special Steel, Non-magnetizable | |
30 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sartorius™ External Adjusting Weight With DKD Calibration Certificate > Mass: 30g ± 0.3mg
Spot an opportunity for improvement?Share a Content Correction