Learn More
Europium(III) oxide, REacton™, 99.996% (REO), Thermo Scientific Chemicals
210.05 € - 1708.50 €
Chemical Identifiers
CAS | 1308-96-9 |
---|---|
Molecular Formula | Eu2O3 |
Molecular Weight (g/mol) | 351.93 |
MDL Number | MFCD00010997 |
InChI Key | AEBZCFFCDTZXHP-UHFFFAOYSA-N |
Synonym | europium oxide, europia, europium trioxide, dieuropium trioxide, europium sesquioxide, europium 3+ oxide, europium oxide euo1.5, europium 3+ ; oxygen 2- |
PubChem CID | 159371 |
IUPAC Name | dieuropium(3+) trioxidandiide |
SMILES | [O--].[O--].[O--].[Eu+3].[Eu+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11458480
|
Thermo Scientific Alfa Aesar
044347.06 |
5 g |
210.05 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11468480
|
Thermo Scientific Alfa Aesar
044347.14 |
25 g |
721.59 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11478480
|
Thermo Scientific Alfa Aesar
044347.22 |
100 g |
1708.50 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Europium(III) oxide is used as an agent for the manufacture of fluorescent glass. It can be used in the anti-counterfeiting phosphors in euro banknotes, red or blue phosphor in television sets. It shows an useful application as an activator for yttrium based phosphors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1308-96-9 | |
351.93 | |
AEBZCFFCDTZXHP-UHFFFAOYSA-N | |
159371 | |
[O--].[O--].[O--].[Eu+3].[Eu+3] |
Eu2O3 | |
MFCD00010997 | |
europium oxide, europia, europium trioxide, dieuropium trioxide, europium sesquioxide, europium 3+ oxide, europium oxide euo1.5, europium 3+ ; oxygen 2- | |
dieuropium(3+) trioxidandiide |
Specifications
2291°C | |
Powder | |
MFCD00010997 | |
europium oxide, europia, europium trioxide, dieuropium trioxide, europium sesquioxide, europium 3+ oxide, europium oxide euo1.5, europium 3+ ; oxygen 2- | |
AEBZCFFCDTZXHP-UHFFFAOYSA-N | |
dieuropium(3+) trioxidandiide | |
159371 | |
99.996% | |
Hygroscopic | |
Europium (III) oxide |
1308-96-9 | |
Eu2O3 | |
14,3904 | |
Insoluble in water. | |
[O--].[O--].[O--].[Eu+3].[Eu+3] | |
351.93 | |
351.92 | |
Reacton™ | |
(REO) |
Safety and Handling
missing translation for 'einecsNumber' : 215-165-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.