missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) chloride hydrate, REacton™, 99.99% (REO), Thermo Scientific Chemicals

Product Code. 11301039
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Europium(III) chloride hydrate is used as a catalyst in organic synthesis. It is involved in the preparation of amino acid based europium(III) complex in order to prepare photoactive thin films, which finds application in organic light emitting diodes. It is also used as a precursor for the preparation of other europium compounds and its salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula EuCl3·xH2O
MDL Number MFCD00010994

Specifications

Color Yellow
Physical Form Crystalline
Quantity 5 g
Assay Percent Range 99.99% (REO)
Solubility Information Soluble in water.
Formula Weight 258.32 (anhydrous)
Odor Odorless
Sensitivity Hygroscopic
Appearance Yellow crystalline
Chemical Name or Material Europium(III) chloride hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'