missing translation for 'onlineSavingsMsg'
Learn More

EDTA disodium salt dihydrate, Reag. Ph. Eur., 99-101%, ≤0.1% NTA (HPLC), for complexometry, Honeywell Fluka™

Reag. Ph. Eur., 99-101%, ≤0.1% NTA (HPLC), for complexometry

25.70 € - 2059.94 €

Chemical Identifiers

CAS 6381-92-6
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number MFCD00150037,MFCD00003541
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
Synonym edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
PubChem CID 44120005
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
View More Specs

Products 8
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15634270
View Documents
Honeywell-Fluka
34549-50G
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15694260
View Documents
Honeywell-Fluka
34549-100G
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15664270
View Documents
Honeywell-Fluka
34549-6X50G
6 x 50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15624270
View Documents
Honeywell-Fluka
34549-500G
500 g
173.81 €
500g
Please to purchase this item. Need a web account? Register with us today!
15644270
View Documents
Honeywell-Fluka
34549-6X100G
6 x 100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15604270
View Documents
Honeywell-Fluka
34549-1KG
1 kg
284.02 €
1kg
Please to purchase this item. Need a web account? Register with us today!
15654270
View Documents
Honeywell-Fluka
34549-6X1KG
6 x 1 kg
1485.35 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15614270
View Documents
Honeywell-Fluka
34549-25KG
25 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

6381-92-6
372.24
OVBJJZOQPCKUOR-UHFFFAOYSA-L
44120005
C10H18N2Na2O10
MFCD00150037,MFCD00003541
edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

Specifications

6381-92-6
MFCD00150037,MFCD00003541
3900609
edta disodium salt, cal-ex decalcifier, buffer solution, ph 10.00, sodium di ethylenediamine tetraacetate dihydrate, ethylenediamine tetraacetic acid, disodium salt dihydrate, ethylenediamine tetraacetic acid, disodium salt, standard solution, sodium di ethylenediamine tetraacetate standard solution, ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
372.24
372.24g/mol
Ethylenediaminetetraacetic acid disodium salt dihydrate
C10H18N2Na2O10
NONH for all modes of transport
>100°C (>212°F)
OVBJJZOQPCKUOR-UHFFFAOYSA-L
disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
44120005
99 to 101%
Videos
Safety and Handling

Safety and Handling

P260

missing translation for 'rtecsNumber' : AH4410000

Product Identifier
  • Ethylenediaminetetraacetic acid disodium salt dihydrate

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications