missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid, Disodium Salt Dihydrate, Certified AR for Analysis, Fisher Chemical™

C10H18N2Na2O10, CAS Number-6381-92-6, 5kg, White, 372.23g/mol, 252 deg.C, 150037, Plastic keg, 4 to 6, Solid

89.05 € - 695.00 €

Chemical Identifiers

CAS 6381-92-6
Molecular Formula C10H18N2Na2O10
Molecular Weight (g/mol) 372.24
MDL Number 150037
InChI Key OVBJJZOQPCKUOR-UHFFFAOYSA-L
IUPAC Name disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
SMILES O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
View More Specs

Products 5
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10335460
View Documents
Fisher Chemical
D/0700/48
100 g Plastic powder jar
90.50 €
100g
Please to purchase this item. Need a web account? Register with us today!
10203570
View Documents
Fisher Chemical
D/0700/50
250 g Plastic powder jar
89.05 €
250g
Please to purchase this item. Need a web account? Register with us today!
10213570
View Documents
Fisher Chemical
D/0700/53
500 g Plastic powder jar
126.00 €
500g
Please to purchase this item. Need a web account? Register with us today!
10526383
View Documents
Fisher Chemical
D/0700/60
1 kg Plastic powder jar
221.00 €
1kg
Please to purchase this item. Need a web account? Register with us today!
11473743
View Documents
Fisher Chemical
D/0700/65
5 kg Plastic keg
695.00 €
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

C10H18N2Na2O10, CAS Number-6381-92-6, 5kg, White, 372.23g/mol, 252 deg.C, 150037, Plastic keg, 4 to 6, Solid

Specifications

Chemical Identifiers

6381-92-6
372.24
OVBJJZOQPCKUOR-UHFFFAOYSA-L
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
C10H18N2Na2O10
150037
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate

Specifications

252°C
White
Solid
150037
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
372.24
6381-92-6
4 to 6
C10H18N2Na2O10
OVBJJZOQPCKUOR-UHFFFAOYSA-L
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
372.23g/mol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Ethylenediaminetetraacetic acid, disodium salt dihydrate

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.

Precautionary Statement
  • P304-IF INHALED:Call a POISON CENTER/doctor if you feelunwell..,IF INHALED:Call a POISON CENTER/doctor if you feelunwell..
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.,Avoid breathing dust/fume/gas/mist/vapours/spray.

Supplemental information
  • MIXTURE LIST-Contains : Ethylenediaminetetraacetic acid, disodium salt dihydrate, Disodium EDTA

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.