missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediamine dihydrochloride, 98%, Thermo Scientific Chemicals

34.50 € - 412.00 €

Chemical Identifiers

CAS 333-18-6
Molecular Formula C2H10Cl2N2
Molecular Weight (g/mol) 133.02
MDL Number MFCD00012524
InChI Key OHHBFEVZJLBKEH-UHFFFAOYSA-N
Synonym ethylenediamine dihydrochloride, chlor-ethamine, ethylenediammonium chloride, ethylenediammonium dichloride, ethane-1,2-diamine dihydrochloride, dimethylenediamine dihydrochloride, 1,2-diaminoethane dihydrochloride, 1,2-ethanediamine, dihydrochloride, ethylenediamine 2hcl, dimethylenediamine diydrochloride
PubChem CID 9521
ChEBI CHEBI:53626
IUPAC Name ethane-1,2-diamine;dihydrochloride
SMILES [H+].[H+].[Cl-].[Cl-].NCCN
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11440064
View Documents
Thermo Scientific Alfa Aesar
A13563.22
100 g
34.50 €
100g
Please to purchase this item. Need a web account? Register with us today!
11450064
View Documents
Thermo Scientific Alfa Aesar
A13563.36
500 g
100.00 €
500g
Please to purchase this item. Need a web account? Register with us today!
11460064
View Documents
Thermo Scientific Alfa Aesar
A13563.0E
2500 g
412.00 €
2500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ethylenediamine dihydrochloride was used in modified ethylenediamine condensation method for fluorimetric determination of catecholamines. It was used to investigate the luminescence properties of complexes of EuIII and TbIII with ethylenediamine. It was used in the synthesis of 1,3,5-tris(4,5-dihydro-1H-imidazol-2-yl)benzene3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylenediamine dihydrochloride was used in modified ethylenediamine condensation method for fluorimetric determination of catecholamines. It was used to investigate the luminescence properties of complexes of EuIII and TbIII with ethylenediamine. It was used in the synthesis of 1,3,5-tris(4,5-dihydro-1H-imidazol-2-yl)benzene3

Solubility
Soluble in water 300 g/L (20°C).

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

333-18-6
133.02
OHHBFEVZJLBKEH-UHFFFAOYSA-N
9521
ethane-1,2-diamine;dihydrochloride
C2H10Cl2N2
MFCD00012524
ethylenediamine dihydrochloride, chlor-ethamine, ethylenediammonium chloride, ethylenediammonium dichloride, ethane-1,2-diamine dihydrochloride, dimethylenediamine dihydrochloride, 1,2-diaminoethane dihydrochloride, 1,2-ethanediamine, dihydrochloride, ethylenediamine 2hcl, dimethylenediamine diydrochloride
CHEBI:53626
[H+].[H+].[Cl-].[Cl-].NCCN

Specifications

333-18-6
C2H10Cl2N2
Hygroscopic
Soluble in water 300g/L (20°C).
[H+].[H+].[Cl-].[Cl-].NCCN
133.02
CHEBI:53626
98%
>300°C
MFCD00012524
ethylenediamine dihydrochloride, chlor-ethamine, ethylenediammonium chloride, ethylenediammonium dichloride, ethane-1,2-diamine dihydrochloride, dimethylenediamine dihydrochloride, 1,2-diaminoethane dihydrochloride, 1,2-ethanediamine, dihydrochloride, ethylenediamine 2hcl, dimethylenediamine diydrochloride
OHHBFEVZJLBKEH-UHFFFAOYSA-N
ethane-1,2-diamine;dihydrochloride
9521
133.02
Ethylenediamine dihydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H361-H373-H302
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.

P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P341-P305+P351+P338-P312-P330-P333+P313-P342+P311-P362-P501c

H302-H315-H317-H319-H334-H335

missing translation for 'einecsNumber' : 206-369-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only