missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene Glycol, Technical, Honeywell™
Technical, suitable as anti-freezing agent (with corrosion-inhibitor), blue
94.36 € - 161.14 €
Chemical Identifiers
CAS | 107-21-1 |
---|---|
Molecular Formula | C2H6O2 |
Molecular Weight (g/mol) | 62.068 |
MDL Number | MFCD00002885 |
InChI Key | LYCAIKOWRPUZTN-UHFFFAOYSA-N |
Synonym | ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol |
PubChem CID | 174 |
ChEBI | CHEBI:30742 |
IUPAC Name | ethane-1,2-diol |
SMILES | C(CO)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15690690
|
Honeywell Research Chemicals
03770-1L |
1 L | Poly Bottle | ||||||
15600700
|
Honeywell Research Chemicals
03770-5L |
5 L | Poly Bottle |
161.14 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Chemicals that do not have an established standard set for quality and impurity levels or for products where the purity is <90%
- Variation in color and physical form is possible
Chemical Identifiers
107-21-1 | |
62.068 | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
174 | |
ethane-1,2-diol |
C2H6O2 | |
MFCD00002885 | |
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol | |
CHEBI:30742 | |
C(CO)O |
Specifications
-20°C | |
Blue | |
195°C to 198°C | |
111°C (231.8°F) | |
Liquid | |
C2H6O2 | |
MFCD00002885 | |
UN3082 | |
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol | |
C(CO)O | |
62.068 | |
CHEBI:30742 | |
Technical |
107-21-1 | |
1.113g/mL at 25°C | |
0.153 | |
752°F | |
n20/D 1.431(lit.) | |
HOCH2CH2OH | |
0.08mmHg (20°C) | |
505945 | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
ethane-1,2-diol | |
174 | |
62.07g/mol | |
Ethylene Glycol |
Safety and Handling
P260-P301 + P312 + P330
H302-H373
missing translation for 'dotInformation' : 9/PGIII
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Ethylene glycol
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Reproductive toxicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H361d-Suspected of damaging the unborn child.
- H373-May cause damage to organs.
Precautionary Statement
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : ethanediol; ethyleneglycol; Sodium 2-ethylhexanoate