missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol-O,O'-bis(2-aminoethyl)-N,N,N',N'-tetraacetic acid, 0.5M aq. soln., pH 8.0, Thermo Scientific Chemicals

100.50 € - 173.87 €

Chemical Identifiers

CAS 67-42-5
Molecular Formula C14H24N2O10
Molecular Weight (g/mol) 380.35
MDL Number MFCD00004291
InChI Key DEFVIWRASFVYLL-UHFFFAOYSA-N
Synonym egta, egtazic acid, gedta, ethylenebis oxyethylenenitrilo tetraacetic acid, ebonta, 6,9-dioxa-3,12-diazatetradecanedioic acid, 3,12-bis carboxymethyl, 1,2-bis 2-bis carboxymethyl amino ethoxy ethane, ethylene glycol tetraacetic acid, h4egta, egtazic acid usan:inn
PubChem CID 6207
ChEBI CHEBI:30740
IUPAC Name 2-[2-[2-[2-[bis(carboxymethyl)amino]ethoxy]ethoxy]ethyl-(carboxymethyl)amino]acetic acid
SMILES C(COCCOCCN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15405795
View Documents Promotion Details
Thermo Scientific Alfa Aesar
J60767.AD
50 mL
100.50 €
50mL
Please to purchase this item. Need a web account? Register with us today!
15415795
View Documents Promotion Details
Thermo Scientific Alfa Aesar
J60767.AE
100 mL
173.87 €
100mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ethylene glycol-O,O'-bis(2-aminoethyl)-N,N,N',N'-tetraacetic acid is a chelating agent used for making complexes, especially for calcium ions, which help determine calcium in the presence of magnesium. It can be used experimentally for treating animals possessing cerium poisoning. It is useful in the separation of thorium from monazite. It also acts as a buffer and is used in protein purification.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylene glycol-O,O′-bis(2-aminoethyl)-N,N,N′,N′-tetraacetic acid is a chelating agent used for making complexes, especially for calcium ions, which help determine calcium in the presence of magnesium. It can be used experimentally for treating animals possessing cerium poisoning. It is useful in the separation of thorium from monazite. It also acts as a buffer and is used in protein purification.

Solubility
Miscible with 0.1M sodium hydroxide solution.

Notes
Incompatible with strong oxidizing agents, alkali, alkali earth metal and many reactive organic and inorganic chemicals.
Specifications

Chemical Identifiers

67-42-5
380.35
DEFVIWRASFVYLL-UHFFFAOYSA-N
6207
2-[2-[2-[2-[bis(carboxymethyl)amino]ethoxy]ethoxy]ethyl-(carboxymethyl)amino]acetic acid
C14H24N2O10
MFCD00004291
egta, egtazic acid, gedta, ethylenebis oxyethylenenitrilo tetraacetic acid, ebonta, 6,9-dioxa-3,12-diazatetradecanedioic acid, 3,12-bis carboxymethyl, 1,2-bis 2-bis carboxymethyl amino ethoxy ethane, ethylene glycol tetraacetic acid, h4egta, egtazic acid usan:inn
CHEBI:30740
C(COCCOCCN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O

Specifications

67-42-5
8.0
MFCD00004291
Miscible with 0.1M sodium hydroxide solution.
C(COCCOCCN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O
380.35
CHEBI:30740
0.5M aq. soln.
Ethylene glycol-O,O'-bis(2-aminoethyl)-N,N,N',N'-tetraacetic acid
Colorless
C14H24N2O10
egta, egtazic acid, gedta, ethylenebis oxyethylenenitrilo tetraacetic acid, ebonta, 6,9-dioxa-3,12-diazatetradecanedioic acid, 3,12-bis carboxymethyl, 1,2-bis 2-bis carboxymethyl amino ethoxy ethane, ethylene glycol tetraacetic acid, h4egta, egtazic acid usan:inn
DEFVIWRASFVYLL-UHFFFAOYSA-N
2-[2-[2-[2-[bis(carboxymethyl)amino]ethoxy]ethoxy]ethyl-(carboxymethyl)amino]acetic acid
6207
380.35
Liquid
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only