missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene glycol, 95%, pure, Thermo Scientific Chemicals
31.40 € - 57.30 €
Chemical Identifiers
CAS | 107-21-1 |
---|---|
Molecular Formula | C2H6O2 |
Molecular Weight (g/mol) | 62.068 |
InChI Key | LYCAIKOWRPUZTN-UHFFFAOYSA-N |
Synonym | ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol |
PubChem CID | 174 |
ChEBI | CHEBI:30742 |
IUPAC Name | ethane-1,2-diol |
SMILES | C(CO)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10107470
|
Thermo Scientific Acros
410010010 |
1 kg | Glass Bottle |
31.40 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11924631
|
Thermo Scientific Acros
410010025 |
2.5 kg | Glass Bottle |
57.30 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
107-21-1 | |
62.068 | |
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol | |
CHEBI:30742 | |
C(CO)O |
C2H6O2 | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
174 | |
ethane-1,2-diol |
Specifications
-13.0°C | |
1.1130g/mL | |
116°C | |
Viscous Liquid | |
95% | |
C2H6O2 | |
21 mPa.s (20°C) | |
ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol | |
LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
ethane-1,2-diol | |
174 | |
62.06 | |
Pure |
107-21-1 | |
196.0°C to 198.0°C | |
Authentic | |
1.4300 to 1.4330 | |
1.113 | |
HOCH2CH2OH | |
01,375; 05,296; 09,217; 15,156 | |
Solubility in water: miscible. Other solubilities: miscible with acetic acid, acetone, glycerol,, pyridine, lower aliphatic alcohols,, aldehydes and ketones, slightly soluble in ether, practically insoluble in benzene and hydrocarbons | |
C(CO)O | |
62.068 | |
CHEBI:30742 | |
95% | |
Ethylene glycol |
Safety and Handling
Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Ethylene glycol
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H373-May cause damage to organs.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
RUO – Research Use Only