missing translation for 'onlineSavingsMsg'
Learn More

Ethyl perfluorooctanoate, 97%, Thermo Scientific™

34.00 € - 142.00 €

Chemical Identifiers

CAS 3108-24-5
Molecular Formula C10H5F15O2
Molecular Weight (g/mol) 442.124
MDL Number MFCD00018063
InChI Key JTOFFHFAQBLPTM-UHFFFAOYSA-N
PubChem CID 76555
IUPAC Name ethyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctanoate
SMILES CCOC(=O)C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11410197
View Documents
Thermo Scientific Alfa Aesar
B21639.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11420197
View Documents
Thermo Scientific Alfa Aesar
B21639.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

3108-24-5
442.124
JTOFFHFAQBLPTM-UHFFFAOYSA-N
ethyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctanoate
C10H5F15O2
MFCD00018063
76555
CCOC(=O)C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

3108-24-5
167°C to 168°C
C10H5F15O2
1.311
1810008
JTOFFHFAQBLPTM-UHFFFAOYSA-N
ethyl 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctanoate
76555
97%
1.626
None
CF3(CF2)6CO2CH2CH3
MFCD00018063
ethyl perfluorooctanoate, ethyl perfluorocaprylate, octanoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-, ethyl ester, octanoic acid,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-, ethyl ester, acmc-20akwv, ethyl perfluorooctanoat, ethyl perfluorooctanonate, octanoic acid, pentadecafluoro-, ethyl ester, pentadecafluorooctanoic acid ethyl ester, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-octanoic acid ethyl ester
CCOC(=O)C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
442.124
442.12
Ethyl perfluorooctanoate
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 221-468-4

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications