missing translation for 'onlineSavingsMsg'
Learn More

Ethyl indole-2-carboxylate, 98+%, Thermo Scientific Chemicals

Product Code. 11370224
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3770-50-1
Molecular Formula C11H11NO2
Molecular Weight (g/mol) 189.21
MDL Number MFCD00005609
InChI Key QQXQAEWRSVZPJM-UHFFFAOYSA-N
Synonym ethyl indole-2-carboxylate, indole-2-carboxylic acid ethyl ester, 2-carbethoxyindole, 1h-indole-2-carboxylic acid, ethyl ester, indole-2-carboxylic acid, ethyl ester, 1h-indole-2-carboxylic acid ethyl ester, ethylindole-2-carboxylate, ethyl 2-indolecarboxylate, 1h-indole-2-carboxylic acid,ethyl ester, 2-carboethoxyindole
PubChem CID 73125
IUPAC Name ethyl 1H-indole-2-carboxylate
SMILES CCOC(=O)C1=CC2=CC=CC=C2N1

Specifications

Melting Point 122°C to 125°C
Quantity 25 g
Beilstein 146395
Formula Weight 189.21
Percent Purity ≥98%
Chemical Name or Material Ethyl indole-2-carboxylate
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.