missing translation for 'onlineSavingsMsg'
Learn More

Ethyl alpha,4-dibromophenylacetate, 97+%, Thermo Scientific Chemicals

Product Code. 11307589
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77143-76-1
Molecular Formula C10H10Br2O2
Molecular Weight (g/mol) 321.996
MDL Number MFCD09953497
InChI Key PPRAWWBOUOBAON-UHFFFAOYSA-N
Synonym ethyl 2-bromo-2-4-bromophenyl acetate, ethyl 2-bromo-4-bromophenyl acetate, ethyl bromo 4-bromophenyl acetate, alpha,4-dibromophenylacetic acid ethyl ester, ethyl alpha,4-dibromophenylacetate, ethyl2-bromo-2-4-bromophenyl acetate, ethyl 2-bromanyl-2-4-bromophenyl ethanoate, benzeneacetic acid, ?,4-dibromo-, ethyl ester, benzeneacetic acid, a,4-dibromo-, ethyl ester
PubChem CID 12754876
IUPAC Name ethyl 2-bromo-2-(4-bromophenyl)acetate
SMILES CCOC(=O)C(C1=CC=C(C=C1)Br)Br

Specifications

Density 1.642
Boiling Point 140°C (5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.571
Quantity 25 g
UN Number UN3265
Solubility Information Slightly soluble in water.
Formula Weight 321.99
Percent Purity ≥97%
Chemical Name or Material Ethyl alpha,4-dibromophenylacetate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'