missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Alcohol, Reagent, Denat., Spectro Grade, Contains 5% 2-PrOH and 5% MeOH, Thermo Scientific Chemicals

45.83 € - 72.56 €

Chemical Identifiers

CAS 64-17-5, 67-56-1, 67-63-0
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10338360
View Documents
Thermo Scientific Acros
224095000
500 mL Glass Bottle
45.83 €
500mL
Please to purchase this item. Need a web account? Register with us today!
10481071
View Documents
Thermo Scientific Acros
224090010
1 L Glass Bottle 72.56 €
1 L
Qty Price Break Available
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for UV/VIS spectroscopy
Specifications

Chemical Identifiers

64-17-5, 67-56-1, 67-63-0
46.069
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
CHEBI:16236
CCO
C2H6O
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
702
ethanol

Specifications

0.01 max. at 300nm, 0.04 max. at 250nm, 0.65 max. at 210nm, 1 max. at 205nm
1.59
0.0005% max.
C2H5OH
0.1% Max. (Coulometric)
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
CCO
46.069
CHEBI:16236
4.0 to 6.0% (2-propanol)
Ethanol
64-17-5, 67-56-1, 67-63-0
Liquid
C2H6O
Vapor Pressure: 59mbar at 20°C
14, 3761
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
702
46.06
Spectroscopy
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Ethanol, denat. with 2-PrOH and MeOH

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H371-May cause damage to organs.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : ethanol, methanol, isopropanol

SDS
Documents

Documents

Product Certifications

For Manufacturing and Laboratory Use. Not for beverage use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.