missing translation for 'onlineSavingsMsg'
Learn More

Ethyl alcohol, Pure 200 Proof (Taxed), Meets USP testing specifications, for domestic sale in Ireland and United Kingdom only, Honeywell Riedel-de Haën™

Product Code. 15635660
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
MDL Number MFCD00003568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -114°C
Absorbance max. 0.10 270-340 nm, max. 0.30 250-260 nm, max. 0.40 at 240 nm
CAS Max % 100.0000%
Density 0.790 - 0.791 g/cm3
Boiling Point 78°C to 79°C
Quantity 1 L
Flash Point 12°C
Packaging Glass Bottle
Linear Formula CH3CH2OH
UN Number UN1170
Beilstein 1718733
Formula Weight 46
Percent Purity >99.5%
Grade Pure
Chemical Name or Material Ethyl alcohol, Pure (Taxed), Meets USP testing specifications, for domestic sale in Germany, France, Ireland and United Kingdom
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'