missing translation for 'onlineSavingsMsg'
Learn More

Ethyl acetoacetate, 99%, pure, Thermo Scientific Chemicals

46.43 € - 399.99 €

Chemical Identifiers

CAS 141-97-9
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
MDL Number MFCD00009199
InChI Key XYIBRDXRRQCHLP-UHFFFAOYSA-N
Synonym ethyl acetoacetate, ethyl acetylacetate, ethyl 3-oxobutyrate, diacetic ether, butanoic acid, 3-oxo-, ethyl ester, ethyl acetylacetonate, acetoacetic acid, ethyl ester, 3-oxobutanoic acid ethyl ester, 3-oxo-butyric acid ethyl ester, active acetylacetate
PubChem CID 8868
ChEBI CHEBI:4893
IUPAC Name ethyl 3-oxobutanoate
SMILES CCOC(=O)CC(=O)C
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10429030
View Documents
Thermo Scientific Acros
117970010
1 L Glass Bottle
46.43 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10162860
View Documents
Thermo Scientific Acros
117970025
2.5 L Glass Bottle
82.41 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10449220
View Documents
Thermo Scientific Acros
117970250
25 L Plastic drum
399.99 €
25 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

141-97-9
130.14
XYIBRDXRRQCHLP-UHFFFAOYSA-N
8868
ethyl 3-oxobutanoate
C6H10O3
MFCD00009199
ethyl acetoacetate, ethyl acetylacetate, ethyl 3-oxobutyrate, diacetic ether, butanoic acid, 3-oxo-, ethyl ester, ethyl acetylacetonate, acetoacetic acid, ethyl ester, 3-oxobutanoic acid ethyl ester, 3-oxo-butyric acid ethyl ester, active acetylacetate
CHEBI:4893
CCOC(=O)CC(=O)C

Specifications

141-97-9
Colorless
180.0°C
Authentic
C6H10O3
1.4180 to 1.4200
03, 632
15, 3812
ethyl acetoacetate, ethyl acetylacetate, ethyl 3-oxobutyrate, diacetic ether, butanoic acid, 3-oxo-, ethyl ester, ethyl acetylacetonate, acetoacetic acid, ethyl ester, 3-oxobutanoic acid ethyl ester, 3-oxo-butyric acid ethyl ester, active acetylacetate
XYIBRDXRRQCHLP-UHFFFAOYSA-N
ethyl 3-oxobutanoate
8868
1.696 mPa.s (20°C)
99%
Liquid
-45.0°C
1.0300g/mL
70°C
98.5% min. (GC)
CH3COCH2CO2C2H5
MFCD00009199
12,222
1.03
Solubility in water: 116g/L (20°C). Other solubilities: soluble in most organic solvent: diethyl ether,,benzene,ethanol,ehyl acetate,chloroform,,acetone
CCOC(=O)CC(=O)C
130.14
CHEBI:4893
130.14
Pure
Ethyl acetoacetate, 99%
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 205-516-1

missing translation for 'rtecsNumber' : AK5250000

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.