missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate 99.8+%, for Residue Analysis, Distol™, Fisher Chemical™

46.80 € - 120.00 €

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number 9171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10783791
View Documents
Fisher Chemical
E/0903/15
1 L
46.80 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10793791
View Documents
Fisher Chemical
E/0903/17
2.5 L
120.00 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

141-78-6
88.106
XEKOWRVHYACXOJ-UHFFFAOYSA-N
CHEBI:27750
CCOC(=O)C
C4H8O2
9171
8857
ethyl acetate

Specifications

141-78-6
Colorless
9171
CCOC(=O)C
88.106
CHEBI:27750
103mbar at 20°C
3.04
75°C
Amber glass bottle
C4H8O2
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl acetate
8857
88.11g/mol
0.45cp at 20°C
-83.5°C
Liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Ethyl acetate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : ethyl acetate

SDS
Documents

Documents

Product Certifications