missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate, puriss., ≥99.5% (GC); Honeywell™ Riedel-de Haën™
BIOPHARMA

Puriss., meets analytical specification of BP, NF, Ph. Eur.,≥99.5% (GC)

286.29 € - 481.37 €

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15652910
BIOPHARMA
View Documents Promotion Details
Honeywell-Riedel-de Haen
27227-6X1L
6 x 1 L Poly Bottle
286.29 €
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15622910
BIOPHARMA
View Documents Promotion Details
Honeywell-Riedel-de Haen
27227-4X2.5L
4 x 2.5 L Poly Bottle
439.84 €
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
15632910
View Documents Promotion Details
Honeywell-Riedel-de Haen
27227-4X5L
4 x 5 L Aluminum Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications

Chemical Identifiers

141-78-6
88.106
XEKOWRVHYACXOJ-UHFFFAOYSA-N
8857
ethyl acetate
C4H8O2
MFCD00009171
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
CHEBI:27750
CCOC(=O)C

Specifications

-84°C
Colorless
76.5°C to 77.5°C
−3°C (26.6°F)
Liquid
CH3COOC2H5
73mmHg (20°C)
506104
XEKOWRVHYACXOJ-UHFFFAOYSA-N
ethyl acetate
8857
88.11g/mol
puriss.
141-78-6
0.898 to 0.902g/mL at 20°C, 0.894 to 0.898g/mL at 25°C
2.2 to 11.5% (38°F)
801°F
C4H8O2
MFCD00009171
UN1173
ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
CCOC(=O)C
88.106
CHEBI:27750
≥99.5% (GC)
Ethyl Acetate, Meets analytical specification of Ph. Eur.BP, NF
Videos
Safety and Handling

Safety and Handling

P210-P305 + P351 + P338-P370 + P378-P403 + P235

H225-H319-H336

missing translation for 'dotInformation' : Class 3 - PG 2 - Ethyl acetate

missing translation for 'einecsNumber' : 205-500-4

missing translation for 'rtecsNumber' : AH5425000

Product Identifier
  • Ethyl acetate

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications