missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Acetate, Extra Pure, SLR, Fisher Chemical™
20.30 € - 329.00 €
Chemical Identifiers
CAS | 141-78-6 |
---|---|
Molecular Formula | C4H8O2 |
Molecular Weight (g/mol) | 88.106 |
MDL Number | 9171 |
InChI Key | XEKOWRVHYACXOJ-UHFFFAOYSA-N |
PubChem CID | 8857 |
ChEBI | CHEBI:27750 |
IUPAC Name | ethyl acetate |
SMILES | CCOC(=O)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11418243
|
Fisher Chemical
E/0850/08 |
500 mL | Amber Glass Bottle |
20.30 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10204340
|
Fisher Chemical
E/0850/15 |
1 L | Amber Glass Bottle |
27.45 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10080130
|
Fisher Chemical
E/0850/17 |
2.5 L | Amber Glass Bottle |
71.55 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11448243
|
Fisher Chemical
E/0850/PC17 |
2.5 L | HDPE Bottle |
69.90 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10409750
|
Fisher Chemical
E/0850/PB17 |
2.5 L | Plastic-coated Glass Bottle |
52.30 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10386370
|
Fisher Chemical
E/0850/21 |
5 L | HDPE Bottle |
92.30 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10409169
|
Fisher Chemical
E/0850/FP21 |
5 L | Plastic Container |
125.00 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10542334
|
Fisher Chemical
E/0850/24 |
10 L | Metal Can |
244.00 €
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10617412
|
Fisher Chemical
E/0850/25 |
25 L | Metal Can |
329.00 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Ethyl Acetate, Extra Pure, SLR
Chemical Identifiers
141-78-6 | |
88.106 | |
XEKOWRVHYACXOJ-UHFFFAOYSA-N | |
CHEBI:27750 | |
CCOC(=O)C |
C4H8O2 | |
9171 | |
8857 | |
ethyl acetate |
Specifications
-83.5°C | |
Colorless | |
Liquid | |
9171 | |
0.45cp at 20°C | |
CCOC(=O)C | |
88.106 | |
CHEBI:27750 |
141-78-6 | |
75°C | |
C4H8O2 | |
103 mbar at 20°C | |
XEKOWRVHYACXOJ-UHFFFAOYSA-N | |
ethyl acetate | |
8857 | |
88.11g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Ethyl acetate
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P240-Ground/bond container and receiving equipment.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.