missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-bromovalerate, 98%, Thermo Scientific Chemicals

Product Code. 11403964
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Ethyl 5-bromovalerate is employed in the preparation of S(γ-Carboxypropyl)-DL-homocysteine and S(δ-Carboxybutyl)-DL-homocysteine. It is also used in the preparation of ethyl 5-azidovalerate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 5-bromovalerate is employed in the preparation of S(γ-Carboxypropyl)-DL-homocysteine and S(δ-Carboxybutyl)-DL-homocysteine. It is also used in the preparation of ethyl 5-azidovalerate.

Solubility
Not miscible or difficult to mix in water.

Notes
Light Sensitive. Keep away from oxidizing agents. Store in dark.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14660-52-7
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.08
MDL Number MFCD00000266
InChI Key AFRWBGJRWRHQOV-UHFFFAOYSA-N
PubChem CID 84580
IUPAC Name ethyl 5-bromopentanoate
SMILES CCOC(=O)CCCCBr

Specifications

Density 1.306
Boiling Point 227°C
Flash Point 103°C (217°F)
Linear Formula Br(CH2)4CO2CH2CH3
Refractive Index 1.458
Quantity 100 g
Beilstein 1752023
Sensitivity Light sensitive
Synonym ethyl 5-bromovalerate, 5-bromopentanoic acid ethyl ester, pentanoic acid, 5-bromo-, ethyl ester, 5-bromovaleric acid ethyl ester, ethyl omega-bromovalerate, ethyl5-bromovalerate, ethyl-5-bromovalerate, pubchem3955, ethyl-5-bromo-valerate, ethyl-5-bromopentanoate
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 209.09
Percent Purity 98%
Chemical Name or Material Ethyl 5-bromovalerate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.