Learn More
Ethyl 5-amino-1,2,3-thiadiazole-4-carboxylate, 98+%, Thermo Scientific Chemicals
132.00 € - 345.00 €
Chemical Identifiers
CAS | 6440-02-4 |
---|---|
Molecular Formula | C5H7N3O2S |
Molecular Weight (g/mol) | 173.19 |
MDL Number | MFCD00778317 |
InChI Key | ZUGOHJWUTNKRMG-UHFFFAOYSA-N |
Synonym | ethyl 5-amino-1,2,3-thiadiazole-4-carboxylate, 5-amino-1,2,3-thiadiazole-4-carboxylic acid ethyl ester, 1,2,3-thiadiazole-4-carboxylic acid, 5-amino-, ethyl ester, 5-amino-1,2,3 thiadiazole-4-carboxylic acid ethyl ester, acmc-20am6n, tos-bb-0200, zugohjwutnkrmg-uhfffaoysa, 5-amino-4-carbethoxy-1,2,3-thiadiazole, 5-amino-4-ethoxycarbonyl-1,2,3-thiadiazole, 1,2,3-thiadiazole-4-carboxylicacid,5-amino-,ethylester |
PubChem CID | 1831000 |
IUPAC Name | ethyl 5-aminothiadiazole-4-carboxylate |
SMILES | CCOC(=O)C1=C(SN=N1)N |
Description
Ethyl 5-amino-1,2,3-thiadiazole-4-carboxylate is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl 5-amino-1,2,3-thiadiazole-4-carboxylate is used as a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
6440-02-4 | |
173.19 | |
ZUGOHJWUTNKRMG-UHFFFAOYSA-N | |
1831000 | |
CCOC(=O)C1=C(SN=N1)N |
C5H7N3O2S | |
MFCD00778317 | |
ethyl 5-amino-1,2,3-thiadiazole-4-carboxylate, 5-amino-1,2,3-thiadiazole-4-carboxylic acid ethyl ester, 1,2,3-thiadiazole-4-carboxylic acid, 5-amino-, ethyl ester, 5-amino-1,2,3 thiadiazole-4-carboxylic acid ethyl ester, acmc-20am6n, tos-bb-0200, zugohjwutnkrmg-uhfffaoysa, 5-amino-4-carbethoxy-1,2,3-thiadiazole, 5-amino-4-ethoxycarbonyl-1,2,3-thiadiazole, 1,2,3-thiadiazole-4-carboxylicacid,5-amino-,ethylester | |
ethyl 5-aminothiadiazole-4-carboxylate |
Specifications
6440-02-4 | |
C5H7N3O2S | |
ethyl 5-amino-1,2,3-thiadiazole-4-carboxylate, 5-amino-1,2,3-thiadiazole-4-carboxylic acid ethyl ester, 1,2,3-thiadiazole-4-carboxylic acid, 5-amino-, ethyl ester, 5-amino-1,2,3 thiadiazole-4-carboxylic acid ethyl ester, acmc-20am6n, tos-bb-0200, zugohjwutnkrmg-uhfffaoysa, 5-amino-4-carbethoxy-1,2,3-thiadiazole, 5-amino-4-ethoxycarbonyl-1,2,3-thiadiazole, 1,2,3-thiadiazole-4-carboxylicacid,5-amino-,ethylester | |
ZUGOHJWUTNKRMG-UHFFFAOYSA-N | |
ethyl 5-aminothiadiazole-4-carboxylate | |
1831000 | |
≥98% |
123°C to 125°C | |
MFCD00778317 | |
Insoluble in water. | |
CCOC(=O)C1=C(SN=N1)N | |
173.19 | |
173.19 | |
Ethyl 5-amino-1,2,3-thiadiazole-4-carboxylate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only