missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 4-formylpyrrole-2-carboxylate, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
Ethyl 4-formylpyrrole-2-carboxylate is a pyrrole building block for proteomics research. Further, it is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl 4-formylpyrrole-2-carboxylate is a pyrrole building block for proteomics research. Further, it is used as a pharmaceutical intermediate.
Solubility
Miscible with ethyl acetate.
Notes
Incompatible with steel and aluminum.
Ethyl 4-formylpyrrole-2-carboxylate is a pyrrole building block for proteomics research. Further, it is used as a pharmaceutical intermediate.
Solubility
Miscible with ethyl acetate.
Notes
Incompatible with steel and aluminum.
Specifications
Specifications
Melting Point | 101°C to 105°C |
Quantity | 250 mg |
Beilstein | 473842 |
Solubility Information | Miscible with ethyl acetate. |
Formula Weight | 167.16 |
Percent Purity | 96% |
Chemical Name or Material | Ethyl 4-formylpyrrole-2-carboxylate |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction