missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (3-fluorobenzoyl)acetate, 98%, Thermo Scientific Chemicals

39.50 € - 384.00 €

Chemical Identifiers

CAS 33166-77-7
Molecular Formula C11H11FO3
Molecular Weight (g/mol) 210.204
MDL Number MFCD03424809
InChI Key MLABEWHVTXMKHP-UHFFFAOYSA-N
PubChem CID 2063904
IUPAC Name ethyl 3-(3-fluorophenyl)-3-oxopropanoate
SMILES CCOC(=O)CC(=O)C1=CC(=CC=C1)F
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15497838
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H61315.MD
250 mg
39.50 €
250mg
Please to purchase this item. Need a web account? Register with us today!
15477838
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H61315.03
1 g
101.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
15487838
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H61315.06
5 g
384.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

33166-77-7
210.204
MLABEWHVTXMKHP-UHFFFAOYSA-N
ethyl 3-(3-fluorophenyl)-3-oxopropanoate
C11H11FO3
MFCD03424809
2063904
CCOC(=O)CC(=O)C1=CC(=CC=C1)F

Specifications

33166-77-7
1.166 g/mL
C11H11FO3
MFCD03424809
MLABEWHVTXMKHP-UHFFFAOYSA-N
ethyl 3-(3-fluorophenyl)-3-oxopropanoate
2063904
98%
Ethyl (3-fluorobenzoyl)acetate
Colorless to Orange
262°C to 263°C
1.517
ethyl 3-3-fluorophenyl-3-oxopropanoate, ethyl 3-fluorobenzoyl acetate, 3-3-fluoro-phenyl-3-oxo-propionic acid ethyl ester, ethyl 3-3-fluoro-phenyl-3-oxopropanoate, benzenepropanoic acid, 3-fluoro-beta-oxo-, ethyl ester, ethyl3-3-fluorophenyl-3-oxopropanoate, acmc-1cl0f, ethyl-m-fluorobenzoylacetate, ethyl 3'-fluorobenzoylacetate, ksc570a6p
CCOC(=O)CC(=O)C1=CC(=CC=C1)F
210.204
210.2
Liquid
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only