Learn More
Ethyl 3,5-dichloro-4-hydroxybenzoate, 99%, Thermo Scientific™
28.60 € - 295.00 €
Chemical Identifiers
CAS | 17302-82-8 |
---|---|
Molecular Formula | C9H8Cl2O3 |
Molecular Weight (g/mol) | 235.06 |
MDL Number | MFCD00016420 |
InChI Key | WMKNGSJJEMFQOT-UHFFFAOYSA-N |
Synonym | 3,5-dichloro-4-hydroxybenzoic acid ethyl ester, benzoic acid, 3,5-dichloro-4-hydroxy-, ethyl ester, 3,5-dichloro-4-hydroxy-benzoic acid ethyl ester, acmc-209e5q, 3-10-00-00363 beilstein handbook reference, wln: qr bg fg dvo2, ethyl 3,5-dichloro-4-hydroxy-benzoate, ethyl 3,5-bis chloranyl-4-oxidanyl-benzoate, benzoic acid,5-dichloro-4-hydroxy-, ethyl ester |
PubChem CID | 28460 |
IUPAC Name | ethyl 3,5-dichloro-4-hydroxybenzoate |
SMILES | CCOC(=O)C1=CC(=C(C(=C1)Cl)O)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11408950
|
Thermo Scientific Alfa Aesar
A12489.06 |
5 g |
28.60 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11474114
|
Thermo Scientific Alfa Aesar
A12489.14 |
25 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11484114
|
Thermo Scientific Alfa Aesar
A12489.22 |
100 g |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsEthyl 3,5-dichloro-4-hydroxybenzoate is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.
Solubility
Soluble in methanol.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
17302-82-8 | |
235.06 | |
WMKNGSJJEMFQOT-UHFFFAOYSA-N | |
28460 | |
CCOC(=O)C1=CC(=C(C(=C1)Cl)O)Cl |
C9H8Cl2O3 | |
MFCD00016420 | |
3,5-dichloro-4-hydroxybenzoic acid ethyl ester, benzoic acid, 3,5-dichloro-4-hydroxy-, ethyl ester, 3,5-dichloro-4-hydroxy-benzoic acid ethyl ester, acmc-209e5q, 3-10-00-00363 beilstein handbook reference, wln: qr bg fg dvo2, ethyl 3,5-dichloro-4-hydroxy-benzoate, ethyl 3,5-bis chloranyl-4-oxidanyl-benzoate, benzoic acid,5-dichloro-4-hydroxy-, ethyl ester | |
ethyl 3,5-dichloro-4-hydroxybenzoate |
Specifications
17302-82-8 | |
C9H8Cl2O3 | |
2649965 | |
Soluble in methanol. | |
CCOC(=O)C1=CC(=C(C(=C1)Cl)O)Cl | |
235.06 | |
235.07 | |
Ethyl 3,5-dichloro-4-hydroxybenzoate |
110°C to 112°C | |
MFCD00016420 | |
3,5-dichloro-4-hydroxybenzoic acid ethyl ester, benzoic acid, 3,5-dichloro-4-hydroxy-, ethyl ester, 3,5-dichloro-4-hydroxy-benzoic acid ethyl ester, acmc-209e5q, 3-10-00-00363 beilstein handbook reference, wln: qr bg fg dvo2, ethyl 3,5-dichloro-4-hydroxy-benzoate, ethyl 3,5-bis chloranyl-4-oxidanyl-benzoate, benzoic acid,5-dichloro-4-hydroxy-, ethyl ester | |
WMKNGSJJEMFQOT-UHFFFAOYSA-N | |
ethyl 3,5-dichloro-4-hydroxybenzoate | |
28460 | |
99% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'einecsNumber' : 241-331-2
missing translation for 'rtecsNumber' : DG7503500
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures