missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-oxocyclopentanecarboxylate, 95+%, Thermo Scientific Chemicals
43.32 € - 357.78 €
Chemical Identifiers
CAS | 611-10-9 |
---|---|
Molecular Formula | C8H12O3 |
Molecular Weight (g/mol) | 156.18 |
MDL Number | MFCD00001412 |
InChI Key | JHZPNBKZPAWCJD-UHFFFAOYSA-N |
PubChem CID | 69136 |
IUPAC Name | ethyl 2-oxocyclopentane-1-carboxylate |
SMILES | CCOC(=O)C1CCCC1=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10772115
|
Thermo Scientific Acros
173580250 |
25 g | Glass bottle |
43.32 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10604672
|
Thermo Scientific Acros
173581000 |
100 g | Glass bottle |
104.52 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10245990
|
Thermo Scientific Acros
173585000 |
500 g | Glass bottle |
357.78 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
611-10-9 | |
156.18 | |
JHZPNBKZPAWCJD-UHFFFAOYSA-N | |
ethyl 2-oxocyclopentane-1-carboxylate |
C8H12O3 | |
MFCD00001412 | |
69136 | |
CCOC(=O)C1CCCC1=O |
Specifications
611-10-9 | |
1.0500g/mL | |
77°C | |
95% min. (GC) | |
(O=)C5H7CO2C2H5 | |
MFCD00001412 | |
1.05 | |
Solubility in water: 3-3.5%. Other solubilities: soluble in most organic solvent | |
CCOC(=O)C1CCCC1=O | |
156.18 | |
156.18 | |
Technical | |
Ethyl 2-oxocyclopentanecarboxylate, tech., 90% |
Colorless to Yellow | |
224.0°C to 228.0°C | |
Authentic | |
C8H12O3 | |
1.4500 to 1.4540 | |
10, 597 | |
ethyl 2-oxocyclopentanecarboxylate, 2-carbethoxycyclopentanone, ethyl cyclopentanone-2-carboxylate, cyclopentanecarboxylic acid, 2-oxo-, ethyl ester, cyclopentanone-2-carboxylic acid ethyl ester, ethyl2-oxocyclopentanecarboxylate, 2-ethoxycarbonyl cyclopentanone, ethyl 2-cyclopentanonecarboxylate, .alpha.-carboethoxy cyclopentanone, ethyl 2-cyclopentanone-1-carboxylate | |
JHZPNBKZPAWCJD-UHFFFAOYSA-N | |
ethyl 2-oxocyclopentane-1-carboxylate | |
69136 | |
95+% | |
Liquid |
Safety and Handling
missing translation for 'einecsNumber' : 210-253-0
Product Identifier
- Ethyl 2-oxocyclopentanecarboxylate
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title