missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2,4-dioxo-4-(2-thienyl)butanoate, 90%, Thermo Scientific™

27.40 € - 138.00 €

Chemical Identifiers

CAS 36983-36-5
Molecular Formula C10H10O4S
Molecular Weight (g/mol) 226.246
MDL Number MFCD00126316
InChI Key GCCFYXYKJZUHMI-UHFFFAOYSA-N
Synonym ethyl 2,4-dioxo-4-thiophen-2-yl butanoate, ethyl 2,4-dioxo-4-2-thienyl butanoate, 2,4-dioxo-4-thiophen-2-yl-butyric acid ethyl ester, 2-thiophenebutanoicacid, a,g-dioxo-, ethyl ester, 2,4-dioxo-4-2-thienyl butanoic acid ethyl ester, 2,4-dioxo-4-thiophen-2-ylbutyric acid ethyl ester
PubChem CID 2799459
IUPAC Name ethyl 2,4-dioxo-4-thiophen-2-ylbutanoate
SMILES CCOC(=O)C(=O)CC(=O)C1=CC=CS1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10059491
View Documents
Thermo Scientific Maybridge
SPB04027DA
1 g Amber Glass Bottle
27.40 €
1g
Please to purchase this item. Need a web account? Register with us today!
10069491
View Documents
Thermo Scientific Maybridge
SPB04027EA
10 g Amber Glass Bottle
138.00 €
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

36983-36-5
226.246
GCCFYXYKJZUHMI-UHFFFAOYSA-N
2799459
CCOC(=O)C(=O)CC(=O)C1=CC=CS1
C10H10O4S
MFCD00126316
ethyl 2,4-dioxo-4-thiophen-2-yl butanoate, ethyl 2,4-dioxo-4-2-thienyl butanoate, 2,4-dioxo-4-thiophen-2-yl-butyric acid ethyl ester, 2-thiophenebutanoicacid, a,g-dioxo-, ethyl ester, 2,4-dioxo-4-2-thienyl butanoic acid ethyl ester, 2,4-dioxo-4-thiophen-2-ylbutyric acid ethyl ester
ethyl 2,4-dioxo-4-thiophen-2-ylbutanoate

Specifications

36983-36-5
90%
MFCD00126316
GCCFYXYKJZUHMI-UHFFFAOYSA-N
ethyl 2,4-dioxo-4-thiophen-2-ylbutanoate
2799459
90%
90.0
C10H10O4S
ethyl 2,4-dioxo-4-thiophen-2-yl butanoate, ethyl 2,4-dioxo-4-2-thienyl butanoate, 2,4-dioxo-4-thiophen-2-yl-butyric acid ethyl ester, 2-thiophenebutanoicacid, a,g-dioxo-, ethyl ester, 2,4-dioxo-4-2-thienyl butanoic acid ethyl ester, 2,4-dioxo-4-thiophen-2-ylbutyric acid ethyl ester
CCOC(=O)C(=O)CC(=O)C1=CC=CS1
226.246
226.25
Ethyl 2, 4-dioxo-4-(2-thienyl)butanoate
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.