missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanolamine, Extra Pure, SLR, Fisher Chemical™
19.80 € - 301.53 €
Chemical Identifiers
CAS | 141-43-5 |
---|---|
Molecular Formula | C2H7NO |
Molecular Weight (g/mol) | 61.08 |
MDL Number | MFCD00008183 |
InChI Key | HZAXFHJVJLSVMW-UHFFFAOYSA-N |
Synonym | ethanolamine, monoethanolamine, aminoethanol, 2-hydroxyethylamine, colamine, glycinol, olamine, 2-amino-1-ethanol, ethanol, 2-amino, ethylolamine |
PubChem CID | 700 |
ChEBI | CHEBI:16000 |
SMILES | NCCO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10508040
|
Fisher Chemical
E/0700/08 |
500 mL | Amber glass bottle | N/A | |||||
10346230
|
Fisher Chemical
E/0700/17 |
2.5 L | Amber glass bottle | N/A | |||||
10287233
|
Fisher Chemical
E/0700/PB17 |
2.5 L | Plastic Coated Glass bottle | N/A | |||||
12618635
|
Fisher Chemical
E/0700/25 |
25 L | Metal can | N/A | |||||
Description
Chemical Identifiers
141-43-5 | |
61.08 | |
HZAXFHJVJLSVMW-UHFFFAOYSA-N | |
700 | |
NCCO |
C2H7NO | |
MFCD00008183 | |
ethanolamine, monoethanolamine, aminoethanol, 2-hydroxyethylamine, colamine, glycinol, olamine, 2-amino-1-ethanol, ethanol, 2-amino, ethylolamine | |
CHEBI:16000 |
Specifications
141-43-5 | |
Colorless | |
170°C | |
MFCD00008183 | |
HZAXFHJVJLSVMW-UHFFFAOYSA-N | |
2-aminoethanol | |
700 | |
CHEBI:16000 | |
0.48mmHg at 20°C |
10°C | |
12 | |
C2H7NO | |
ethanolamine, monoethanolamine, aminoethanol, 2-hydroxyethylamine, colamine, glycinol, olamine, 2-amino-1-ethanol, ethanol, 2-amino, ethylolamine | |
NCCO | |
61.08 | |
2.1 | |
61.08g/mol | |
Liquid |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Ethanolamine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/A§.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title