missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol, purum, 96%, Honeywell™
247.39 € - 4099.75 €
Chemical Identifiers
CAS | 64-17-5 |
---|---|
Molecular Formula | C2H6O |
Molecular Weight (g/mol) | 46.069 |
MDL Number | MFCD00003568 |
InChI Key | LFQSCWFLJHTTHZ-UHFFFAOYSA-N |
Synonym | ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol |
PubChem CID | 702 |
ChEBI | CHEBI:16236 |
IUPAC Name | ethanol |
SMILES | CCO |
Description
Chemical Identifiers
64-17-5 | |
46.069 | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
702 | |
ethanol |
C2H6O | |
MFCD00003568 | |
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol | |
CHEBI:16236 | |
CCO |
Specifications
64-17-5 | |
CH3CH2OH | |
UN1170 | |
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol | |
CCO | |
46.069 | |
CHEBI:16236 | |
96% | |
0.789g/mL at 25°C | |
14°C (57.2°F) | |
Fine Spirit | |
Ethanol |
C2H6O | |
MFCD00003568 | |
1718733 | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
ethanol | |
702 | |
46.07g/mol | |
purum | |
19% (60°F) | |
683°F | |
44.6mmHg (20°C) |
Safety and Handling
P210-P305 + P351 + P338-P370 + P378-P403 + P235
H225-H319
missing translation for 'dotInformation' : class 3 - PG 2 - Ethanol
missing translation for 'einecsNumber' : 200-578-6
missing translation for 'rtecsNumber' : KQ6300000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- ethanol
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P264-Wash skin thoroughly after handling.
- P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P233-Keep container tightly closed.
- P243-Take precautionary measures against static discharge.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P240-Ground/bond container and receiving equipment.
- P242-Use only non-sparking tools.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : ethanol