missing translation for 'onlineSavingsMsg'
Learn More

Ethanol (Untaxed), Puriss., meets analytical spec. of BP, Ph. Eur., 96% (v/v), for license holders only, Honeywell Riedel-de Haën™

Product Code. 15602490
Click to view available options
Quantity:
1 L
2.5 L
5 L
30 kg
4 x 2.5 L
4 x 5 L
160 kg
Packaging:
Glass Bottle
Metal Drum
Plastic Bottle
Poly Bottle
Stainless Steel Can
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -114°C (literature)
CAS Max % 100.0000%
Linear Formula NaHSO3
MDL Number MFCD00003530
UN Number UN1170
Beilstein 1718733
Formula Weight 46
Percent Purity 96%
Grade puriss.
Density 0.789 g/mL at 25 °C(lit.)
Boiling Point 78°C to 79°C
Quantity 160 kg
Flash Point 12°C
Packaging Metal Drum
Chemical Name or Material Ethanol (Untaxed), Puriss., meets analytical specification of BP, Ph. Eur., 96% (v/v), for license holders only
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'