missing translation for 'onlineSavingsMsg'
Learn More

Alla France™ Ethanol Hydrometer

Specifications

Quantity 1Each
Type Ethanol Hydrometer
Accuracy ±0.1%
Length (Metric) 350 mm
View More Specs

Description

Description

Specifications

Specifications

1Each
Ethanol Hydrometer
±0.1%
350 mm
Videos
SDS
Documents

Documents

Product Certifications