missing translation for 'onlineSavingsMsg'
Learn More

Ethanol, CHROMASOLV™ ACS Reagent, Honeywell Riedel-de Haën™

Product Code. 15670590
Click to view available options
Quantity:
500 mL
2.5 L
6 x 500 mL
4 x 2.5 L
6 x 1 L
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
MDL Number MFCD00003568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Density 0.789g/mL at 25°C
Quantity 6 x 500 mL
Explosion Limit 19% (60°F)
Flash Point 14°C (57.2°F)
Ignition Point 683°F
Physical Form Extra Fine Spirit
Linear Formula CH3CH2OH
Vapor Pressure 44.6mmHg (20°C)
UN Number 1170
Beilstein 1718733
Formula Weight 46.07g/mol
Percent Purity 96% (v/v)
Grade CHROMASOLV™ ACS Reagent
Chemical Name or Material Ethanol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.