missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol, Absolute, Primar™ for Trace Elemental Analysis, Fisher Chemical
Click to view available options
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Specifications
Specifications
pH | 7 |
Melting Point | -114°C |
CAS | 64-17-5 |
Color | Colorless |
Boiling Point | 78°C |
Quantity | 2.5 L |
Packaging | Amber Glass Bottle |
Physical Form | Liquid |
Molecular Formula | C2H6O |
MDL Number | 3568 |
Show More |
Safety and Handling
Product Identifier
- Ethanol, absolute
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction