missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol Absolute, 99.9%, BAKER ANALYZED™, J.T. Baker™
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
HDPE Bottle
Description
BAKER ANALYZED™ reagents are a grade of high quality chemicals for laboratory and specialized industrial use. Every BAKER ANALYZED™ Reagent label provides an Actual Lot Analysis as a guarantee of purity and consistency. Analytical methods used for these determinations are based on standard test procedures described in technical publications.
Specifications
Specifications
Melting Point | −114.5°C |
Color | Colorless |
Percent Purity | Min. 99.9% v/v |
Density | 0.79g/cm3 at 20°C |
Boiling Point | 78°C |
Quantity | 2.5 L |
Explosion Limit | Lower: 3.5 Vol %, Upper: 15 Vol % |
Flash Point | 12°C |
Ignition Point | 425°C |
Odor | Alcohol |
Show More |
Safety and Handling
Product Identifier
- Ethanol Absolute
- Danger
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P240-Ground/bond container and receiving equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'