missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol, 96% (v/v), Extra Pure, SLR, Fisher Chemical™
142.00 € - 353.00 €
Chemical Identifiers
CAS | 64-17-5 |
---|---|
Molecular Formula | C2H6O |
Molecular Weight (g/mol) | 46.07 |
MDL Number | 3568 |
InChI Key | LFQSCWFLJHTTHZ-UHFFFAOYSA-N |
PubChem CID | 702 |
ChEBI | CHEBI:16236 |
IUPAC Name | ethanol |
SMILES | CCO |
Product Code | Brand | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging | Price | Quantity & Availability | |||||
10162252
|
Fisher Chemical
E/0551DF/17 |
Amber Glass Bottle |
353.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10253912
|
Fisher Chemical
E/0551DF/C17 |
HDPE Bottle |
142.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
64-17-5 | |
46.07 | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
CHEBI:16236 | |
CCO |
C2H6O | |
3568 | |
702 | |
ethanol |
Specifications
-114°C | |
Colorless | |
2.5 L | |
C2H6O | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
ethanol | |
702 | |
46.07g/mol |
64-17-5 | |
78°C | |
Liquid | |
3568 | |
CCO | |
46.07 | |
CHEBI:16236 |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Ethanol (96% v/v)
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title