Learn More
Thermo Scientific™ ES Series FMS Lab Freezers
The new Thermo Scientific™ ES Series FMS Lab Freezers are ideal for storing potentially flammable material in general laboratory settings. Household and commercial freezers are not suitable for such material: they have components in their electrical and refrigeration systems that can trigger explosions of flammable air-vapor mixtures inside the unit.
Description
Description
Reduce risk of accidental explosion and associated sample damage. The new Thermo Scientific™ ES Series FMS Lab Freezers are ideal for storing potentially flammable material in general laboratory settings. Household and commercial freezers are not suitable for such material: they have components in their electrical and refrigeration systems that can trigger explosions of flammable air-vapor mixtures inside the unit. These freezers have been constructed without components that could trigger an explosion or fire of the materials inside.
  • Integrated controller
  • Digital display
  • High- and low-temperature alarms
  • Door ajar alarm
  • Remote alarm contacts
  • Standard door locks
  • Access ports
  • CE/ATEX marked
  • Reversible doors
  • Hydrocarbn refrigerants
  • HCFC and CFC free insulation
Ordering Information: Available in 50Hz regions only; not available in North America and other 60Hz regions.
    Warranty varies by country, please contact us for details
Videos
SDS
Documents
Documents
Product Certifications
FMS units are not designed for use in Class I, Zone 0 or Zone 1 applications which require an explosion-proof unit.
Product Content Correction
Product Title
Thermo Scientific™ ES Series FMS Lab Freezers