Learn More
Tocris Bioscience™ ERK5-IN-1
Potent and selective ERK5 inhibitor
Brand:  Tocris Bioscience™ 5393/50
1115.30 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18732988
  • 1255.00 € / 50mg
Explore more special offers
Description
Description
ERK5-IN-1 is a potent and selective ERK5 inhibitor; inhibits EGFR-induced ERK5 autophosphorylation (EC50 = 90 nM) and ERK5 enzymatic activity (IC50 = 162 nM). Exhibits at least 30-fold selectivity for ERK5 over LRRK2 in a cell based assay. Also selective for ERK5 over a panel of other kinases. Orally bioavailable.
Specifications
Specifications
>98%
638.8
50 mg
ERK5-IN-1
ERK5-IN-1