Learn More
Ergocalciferol, 97.0 to 103.0%, Thermo Scientific™
150.00 € - 1907.00 €
Chemical Identifiers
CAS | 50-14-6 |
---|---|
Molecular Formula | C28H44O |
Molecular Weight (g/mol) | 396.66 |
MDL Number | MFCD00166988,MFCD11656674 |
InChI Key | MECHNRXZTMCUDQ-VLOQVYPSSA-N |
Synonym | ergocalciferol, vitamin d2, calciferol, viosterol, oleovitamin d2, ercalciol, ergorone, detalup, condocaps, crystallina |
PubChem CID | 134129658 |
IUPAC Name | (1S,3E)-3-{2-[(1R,3aS,4E,7aR)-1-[(2R,3E,5R)-5,6-dimethylhept-3-en-2-yl]-7a-methyl-octahydro-1H-inden-4-ylidene]ethylidene}-4-methylidenecyclohexan-1-ol |
SMILES | CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C1/C[C@@H](O)CCC1=C |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
50-14-6 | |
396.66 | |
MECHNRXZTMCUDQ-VLOQVYPSSA-N | |
134129658 | |
CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C1/C[C@@H](O)CCC1=C |
C28H44O | |
MFCD00166988,MFCD11656674 | |
ergocalciferol, vitamin d2, calciferol, viosterol, oleovitamin d2, ercalciol, ergorone, detalup, condocaps, crystallina | |
(1S,3E)-3-{2-[(1R,3aS,4E,7aR)-1-[(2R,3E,5R)-5,6-dimethylhept-3-en-2-yl]-7a-methyl-octahydro-1H-inden-4-ylidene]ethylidene}-4-methylidenecyclohexan-1-ol |
Specifications
50-14-6 | |
C28H44O | |
UN2811 | |
ergocalciferol, vitamin d2, calciferol, viosterol, oleovitamin d2, ercalciol, ergorone, detalup, condocaps, crystallina | |
CC(C)[C@@H](C)\C=C\[C@@H](C)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C1/C[C@@H](O)CCC1=C | |
396.66 | |
396.65 | |
Powder |
118°C to 119°C | |
MFCD00166988,MFCD11656674 | |
1916682 | |
MECHNRXZTMCUDQ-VLOQVYPSSA-N | |
(1S,3E)-3-{2-[(1R,3aS,4E,7aR)-1-[(2R,3E,5R)-5,6-dimethylhept-3-en-2-yl]-7a-methyl-octahydro-1H-inden-4-ylidene]ethylidene}-4-methylidenecyclohexan-1-ol | |
134129658 | |
97.0 to 103.0% | |
Ergocalciferol |
Safety and Handling
P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P310-P312-P314-P330-P361-P363-P501c
H301+H311-H330-H372
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: II
missing translation for 'einecsNumber' : 200-014-9
missing translation for 'rtecsNumber' : KE1050000
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold