Learn More
Erbium(III) chloride, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals
187.94 € - 1434.00 €
Chemical Identifiers
CAS | 10138-41-7 |
---|---|
Molecular Formula | Cl3Er |
Molecular Weight (g/mol) | 273.609 |
MDL Number | MFCD00010989 |
InChI Key | HDGGAKOVUDZYES-UHFFFAOYSA-K |
Synonym | erbium chloride, erbium trichloride, erbium chloride ercl3, ercl3, erbium iii chloride, anhydrous, erbium iii chloride, ultra dry, erbium iii chloride hexahydrate, erbium iii chloride, anhydrous, ?ercl3, erbium iii chloride, anhydrous, powder trace metals basis, erbium iii chloride, anhydrous, beads,-10 mesh trace metals basis |
PubChem CID | 66277 |
IUPAC Name | trichloroerbium |
SMILES | Cl[Er](Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
11433860
|
Thermo Scientific Alfa Aesar
089917.09 |
10 g |
187.94 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11443860
|
Thermo Scientific Alfa Aesar
089917.18 |
50 g |
805.01 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11346409
|
Thermo Scientific Alfa Aesar
089917.30 |
250 g | N/A | N/A | N/A | |||||
Description
It is a starting material for new edge-bridged octahedral M6 cluster compounds which are used for electron and stability studies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10138-41-7 | |
273.609 | |
HDGGAKOVUDZYES-UHFFFAOYSA-K | |
66277 | |
Cl[Er](Cl)Cl |
Cl3Er | |
MFCD00010989 | |
erbium chloride, erbium trichloride, erbium chloride ercl3, ercl3, erbium iii chloride, anhydrous, erbium iii chloride, ultra dry, erbium iii chloride hexahydrate, erbium iii chloride, anhydrous, ?ercl3, erbium iii chloride, anhydrous, powder trace metals basis, erbium iii chloride, anhydrous, beads,-10 mesh trace metals basis | |
trichloroerbium |
Specifications
1500°C | |
10138-41-7 | |
Cl3Er | |
erbium chloride, erbium trichloride, erbium chloride ercl3, ercl3, erbium iii chloride, anhydrous, erbium iii chloride, ultra dry, erbium iii chloride hexahydrate, erbium iii chloride, anhydrous, ?ercl3, erbium iii chloride, anhydrous, powder trace metals basis, erbium iii chloride, anhydrous, beads,-10 mesh trace metals basis | |
HDGGAKOVUDZYES-UHFFFAOYSA-K | |
trichloroerbium | |
66277 | |
99.9% | |
Hygroscopic | |
Erbium (III) chloride, Anhydrous |
774°C | |
Beads | |
MFCD00010989 | |
Soluble in water,strong mineral acids. Slightly soluble in MeOH and THF. | |
Cl[Er](Cl)Cl | |
273.609 | |
273.62 | |
Odorless | |
(metals basis) |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'einecsNumber' : 233-385-0
missing translation for 'rtecsNumber' : KD8575000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.